Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010009.hg.1 |
ENST00000369295.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010009.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010009.hg.1 |
TC10000829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161345.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161345.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161345.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161345.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161345.hg.1 |
TC1000008968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161346.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161346.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161346.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161346.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161346.hg.1 |
TC1000008968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
ENST00000369295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008968.hg.1 |
NM_000684.2 |
Refseq |