Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046186.hg.1 |
ENST00000369771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046186.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046186.hg.1 |
TC01002974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046188.hg.1 |
ENST00000369771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046188.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046188.hg.1 |
TC01002974.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015243.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213630.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213630.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213630.hg.1 |
TC0100015243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213630.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213630.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213631.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213631.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213631.hg.1 |
TC0100015243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213631.hg.1 |
ENST00000369771.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213631.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213633.hg.1 |
NM_005549.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213633.hg.1 |
TC0100015243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213633.hg.1 |
NM_005549.2 |
Refseq |