Assay Details
Gene Symbol: PRO0628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC015320.1 | - | 1 | 212 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3324137 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3863 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3864 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37087 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3324137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816602 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816616 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168757 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816616 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816602 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168706 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168761 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168757 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168740 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168728 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168699 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11168746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31816582 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168699 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168750 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168711 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168739 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168767 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168743 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168745 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168716 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168721 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168705 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168762 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168723 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168709 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168774 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168756 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168770 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168741 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168714 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168726 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168733 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168694 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17192387 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168719 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17192380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168776 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168702 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168778 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168698 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168725 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168729 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168754 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168737 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168700 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168735 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11168707 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17192384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Human Genome U95 Set | 87367_PM_r_at | - | - |
Human Genome U95 Set | 87367_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2213834 | BC015320.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2213834 | BC015320.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2213834 | BC015320.1 | human_transcriptome |
