Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199742.1 | NP_001186671.1 | 1 - 2 | 360 | 110 | |
NM_001924.3 | NP_001915.1 | 1 - 2 | 360 | 110 | ||
GenBank mRNA | AK314991.1 | - | 1 - 2 | 360 | 110 | |
BC011757.2 | - | 1 - 2 | 323 | 3530326 | 110 | |
EF581092.1 | - | 1 - 2 | 323 | 110 | ||
M60974.1 | - | 1 - 2 | 325 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000745.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314991.1, BC011757.2, EF581092.1, M60974.1, NM_001199742.1, NM_001924.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
G2/M transition of mitotic cell cycle
DNA repair
negative regulation of protein kinase activity
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
centrosome cycle
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of JNK cascade
regulation of cell cycle
cellular response to mechanical stimulus
cellular response to ionizing radiation
positive regulation of p38MAPK cascade
positive regulation of reactive oxygen species metabolic process
core promoter sequence-specific DNA binding
protein binding
regulation of cyclin-dependent protein serine/threonine kinase activity
G2/M transition of mitotic cell cycle
DNA repair
negative regulation of protein kinase activity
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
centrosome cycle
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of JNK cascade
regulation of cell cycle
cellular response to mechanical stimulus
cellular response to ionizing radiation
positive regulation of p38MAPK cascade
positive regulation of reactive oxygen species metabolic process
core promoter sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011959.hg.1 | ENST00000370986.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011959.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011959.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011959.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011959.hg.1 | TC01000745.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370985.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008664.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | NM_001924.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | TC0100008664.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000370986.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000460575.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000484245.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | ENST00000617962.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | NM_001199742.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100161645.hg.1 | NM_001924.3 | Refseq |
