Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001924.3 | NP_001915.1 | 2 - 3 | 464 | 77 | |
GenBank mRNA | AK314991.1 | - | 2 - 3 | 464 | 77 | |
BC011757.2 | - | 2 - 3 | 427 | 3530326 | 77 | |
M60974.1 | - | 2 - 3 | 429 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
