Assay Details
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U68030.1 | - | 1 - 2 | 62 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U68030.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000341935.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000349984.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | ENST00000400926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_004367.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014204.hg.1 | NM_031409.3 | Refseq |
