Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002344.hg.1 |
ENST00000376105.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002344.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002344.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002344.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002344.hg.1 |
TC20000194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002342.hg.1 |
ENST00000376105.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002342.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002342.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002342.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002342.hg.1 |
TC20000194.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
TC2000007083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148341.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
TC2000007083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148345.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
TC2000007083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148340.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
TC2000007083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148344.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376105.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
ENST00000376112.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_002165.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007083.hg.1 |
NM_181353.2 |
Refseq |