Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201269.2 | NP_958357.1 | 3 - 4 | 3368 | 89 | |
XM_005271257.4 | XP_005271314.1 | 4 - 5 | 3704 | 89 | ||
XM_005271260.4 | XP_005271317.1 | 3 - 4 | 3681 | 89 | ||
XM_011542258.2 | XP_011540560.1 | 3 - 4 | 3382 | 89 | ||
XM_011542259.2 | XP_011540561.1 | 5 - 6 | 4058 | 89 | ||
XM_011542260.2 | XP_011540562.1 | 5 - 6 | 3962 | 89 | ||
XM_011542261.2 | XP_011540563.1 | 3 - 4 | 3303 | 89 | ||
XM_017002487.1 | XP_016857976.1 | 4 - 5 | 3493 | 89 | ||
XM_017002488.1 | XP_016857977.1 | 4 - 5 | 3704 | 89 | ||
XM_017002489.1 | XP_016857978.1 | 3 - 4 | 3679 | 89 | ||
XM_017002490.1 | XP_016857979.1 | 4 - 5 | 3491 | 89 | ||
XM_017002491.1 | XP_016857980.1 | 3 - 4 | 3303 | 89 | ||
GenBank mRNA | AB019255.1 | - | 3107 | 89 | ||
AB033047.1 | - | 1 - 2 | 2951 | 89 | ||
AF208847.1 | - | 1 - 2 | 1851 | 89 | ||
AK001587.1 | - | 1 - 2 | 1133 | 89 | ||
AK023596.1 | - | 1 - 2 | 307 | 89 | ||
BC063683.1 | - | 2 - 3 | 3062 | 89 | ||
BC132776.1 | - | 3 - 4 | 3183 | 40146798 | 89 | |
BC132778.1 | - | 3 - 4 | 3183 | 40146800 | 89 | |
BC150177.1 | - | 3 - 4 | 3323 | 40148078 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
