Assay Details
Gene Symbol: BNIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY078984.1 | - | 4 - 5 | 602 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000368931.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000392802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000485855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000491386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_001159642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | NM_138278.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000295294.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009936.hg.1 | ENST00000361277.9 | Ensembl |
