Assay Details
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054686.1 | - | 4 - 5 | 572 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
