Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174921.2 | NP_777581.1 | 1 - 2 | 265 | 65 | |
GenBank mRNA | AK023351.1 | - | 1 - 2 | 128 | 65 | |
AK307146.1 | - | 1 - 2 | 158 | 65 | ||
BC008502.1 | - | 1 - 2 | 128 | 4295121 | 65 | |
HY155982.1 | - | 1 - 2 | 266 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001132.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC008502.1, NM_174921.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015533.hg.1 | ENST00000295958.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015533.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015533.hg.1 | ENST00000511809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015533.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015533.hg.1 | TC04001132.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04015538.hg.1 | ENST00000295958.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015538.hg.1 | ENST00000511809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015538.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015538.hg.1 | TC04001132.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0700010663.hg.1 | ENST00000438721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015532.hg.1 | ENST00000295958.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015532.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015532.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015532.hg.1 | TC04001132.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04015535.hg.1 | ENST00000295958.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015535.hg.1 | ENST00000511809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015535.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015535.hg.1 | TC04001132.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | ENST00000630374.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010470.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | TC0400010470.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171517.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | TC0400010470.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171520.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | TC0400010470.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000510628.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | ENST00000512441.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171514.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | TC0400010470.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000295958.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000505729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000507613.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | ENST00000511809.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171516.hg.1 | NM_174921.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010663.hg.1 | ENST00000438721.1 | Ensembl |
