Assay Details
Gene Symbol: FBXW8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012174.1 | NP_036306.1 | 3 - 4 | 476 | 111 | |
NM_153348.2 | NP_699179.2 | 3 - 4 | 674 | 111 | ||
XM_017019175.1 | XP_016874664.1 | 3 - 4 | 451 | 111 | ||
XM_017019176.1 | XP_016874665.1 | 4 - 5 | 683 | 111 | ||
XM_017019177.1 | XP_016874666.1 | 4 - 5 | 683 | 111 | ||
GenBank mRNA | AF176707.1 | - | 3 - 4 | 454 | 111 | |
AK289621.1 | - | 3 - 4 | 467 | 111 | ||
BC037296.2 | - | 3 - 4 | 674 | 4821240 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000920.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | ENST00000309909.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | ENST00000505227.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011963.hg.1 | TC12000920.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | TC1200009036.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200166908.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000309909.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000455858.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | ENST00000551773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_012174.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009036.hg.1 | NM_153348.2 | Refseq |
