Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000551773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
ENST00000551773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
ENST00000309909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
ENST00000505227.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011970.hg.1 |
TC12000920.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
TC1200009036.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166917.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000309909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000455858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000551773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
ENST00000551773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_012174.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009036.hg.1 |
NM_153348.2 |
Refseq |