Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
ENST00000372599.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
ENST00000372600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008698.hg.1 |
TC09000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008697.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008697.hg.1 |
ENST00000372600.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008697.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008697.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008697.hg.1 |
TC09000704.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000483638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000483638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008694.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008694.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008694.hg.1 |
TC09000704.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
TC0900008891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159954.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
TC0900008891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159956.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372599.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000372600.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000483638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000483638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
ENST00000492784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_001127245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008891.hg.1 |
NM_019594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159951.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159951.hg.1 |
NM_001127244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159951.hg.1 |
TC0900008891.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900159951.hg.1 |
ENST00000259324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159951.hg.1 |
NM_001127244.1 |
Refseq |