Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 22 - 23 | 4142 | 61 | |
XM_005270004.3 | XP_005270061.1 | 22 - 23 | 4257 | 61 | ||
XM_011539986.2 | XP_011538288.2 | 22 - 23 | 4260 | 61 | ||
XM_011539987.2 | XP_011538289.2 | 22 - 23 | 4263 | 61 | ||
XM_011539988.2 | XP_011538290.1 | 23 - 24 | 4341 | 61 | ||
XM_011539990.2 | XP_011538292.1 | 22 - 23 | 4288 | 61 | ||
XM_011539991.2 | XP_011538293.1 | 22 - 23 | 4292 | 61 | ||
XM_011539992.2 | XP_011538294.1 | 22 - 23 | 4292 | 61 | ||
XM_017016463.1 | XP_016871952.1 | 23 - 24 | 4184 | 61 | ||
XM_017016464.1 | XP_016871953.1 | 4 - 5 | 571 | 61 | ||
XM_017016465.1 | XP_016871954.1 | 22 - 23 | 4269 | 61 | ||
GenBank mRNA | AK074085.1 | - | 2 - 3 | 200 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
