Assay Details
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020945.1 | NP_065996.1 | 37 - 38 | 6413 | 81 | |
XM_005270004.3 | XP_005270061.1 | 37 - 38 | 6528 | 81 | ||
XM_011539986.2 | XP_011538288.2 | 37 - 38 | 6531 | 81 | ||
XM_011539987.2 | XP_011538289.2 | 37 - 38 | 6534 | 81 | ||
XM_011539988.2 | XP_011538290.1 | 38 - 39 | 6612 | 81 | ||
XM_011539990.2 | XP_011538292.1 | 37 - 38 | 6559 | 81 | ||
XM_017016463.1 | XP_016871952.1 | 38 - 39 | 6455 | 81 | ||
XM_017016464.1 | XP_016871953.1 | 19 - 20 | 2842 | 81 | ||
GenBank mRNA | AB046827.1 | - | 4 - 5 | 596 | 81 | |
AK074085.1 | - | 17 - 18 | 2471 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000323.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
