Assay Details
Gene Symbol: NPY6R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D86519.1 | - | 1 | 49 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heterogeneous nuclear ribonucleoprotein A0 | HNRNPA0 | HNRPA0 | -53.4kb | Anti-sense | Chr. 5 - 137751384 - 137754350 on NCBI Build GRCh38 |
Downstream | myotilin | MYOT |
LGMD1, LGMD1A, MFM3, TTID, TTOD |
58.1kb | Sense | Chr. 5 - 137867282 - 137887851 on NCBI Build GRCh38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D86519.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
