Assay Details
Gene Symbol: CDC42BPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017525.2 | NP_059995.2 | 4 - 5 | 428 | 65 | |
XM_011545155.2 | XP_011543457.1 | 4 - 5 | 543 | 65 | ||
XM_011545156.2 | XP_011543458.1 | 4 - 5 | 544 | 65 | ||
XM_011545157.2 | XP_011543459.1 | 4 - 5 | 544 | 65 | ||
XM_011545158.2 | XP_011543460.1 | 4 - 5 | 544 | 65 | ||
XM_011545159.2 | XP_011543461.1 | 4 - 5 | 544 | 65 | ||
XM_011545160.2 | XP_011543462.1 | 5 - 6 | 495 | 65 | ||
XM_011545161.2 | XP_011543463.1 | 4 - 5 | 544 | 65 | ||
XM_017017996.1 | XP_016873485.1 | 4 - 5 | 543 | 65 | ||
XM_017017997.1 | XP_016873486.1 | 4 - 5 | 544 | 65 | ||
XM_017017998.1 | XP_016873487.1 | 4 - 5 | 544 | 65 | ||
GenBank mRNA | AY648038.1 | - | 4 - 5 | 428 | 65 | |
Y12337.1 | - | 1 - 2 | 92 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001914.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11022857.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11022857.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11022857.hg.1 | TC11001914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100185552.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100185552.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100185552.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100185552.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100185552.hg.1 | TC1100011188.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000342711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000468512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000480767.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | ENST00000491280.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011188.hg.1 | NM_017525.2 | Refseq |
