Assay Details
Gene Symbol: CDC42EP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145057.3 | NP_659494.2 | 2 - 3 | 373 | 132 | |
GenBank mRNA | BC024327.1 | - | 2 - 3 | 344 | 4390710 | 132 |
BC065910.1 | - | 2 - 3 | 278 | 6572564 | 132 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001849.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC065910.1,NM_145057.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
