Assay Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC056256.1 | - | 3 - 4 | 418 | 63 | |
BC070334.1 | - | 3 - 4 | 413 | 63 | ||
BC073791.1 | - | 3 - 4 | 404 | 63 | ||
BC095489.1 | - | 3 - 4 | 408 | 63 |
Gene Symbol: IGKV1-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC030814.1 | - | 3 - 4 | 427 | 63 | |
BC034146.1 | - | 3 - 4 | 413 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
