Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156370.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156370.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156370.hg.1 |
TC1300008351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156370.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156370.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156371.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156371.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156371.hg.1 |
TC1300008351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156371.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156371.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156372.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156372.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156372.hg.1 |
TC1300008351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156372.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156372.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
ENST00000430733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008351.hg.1 |
NR_046531.1 |
Refseq |