Assay Details
Gene Symbol: LOC101927865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110283.1 | 1 - 2 | 134 | 72 | ||
GenBank mRNA | AI925949.1 | - | 2 - 3 | 112 | 72 | |
HY006998.1 | - | 1 - 2 | 135 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000430494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010595.hg.1 | ENST00000440326.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000417096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000432413.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000440545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | ENST00000448786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016765.hg.1 | NR_110283.1 | Refseq |
