Assay Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198542.1 | NP_001185471.1 | 4 - 5 | 491 | 78 | |
NM_001198543.1 | NP_001185472.1 | 4 - 5 | 461 | 78 | ||
NM_001270974.2 | NP_001257903.1 | 4 - 5 | 559 | 78 | ||
NM_017558.4 | NP_060028.2 | 4 - 5 | 559 | 78 | ||
XM_006721206.2 | XP_006721269.1 | 4 - 5 | 453 | 78 | ||
XM_011523146.2 | XP_011521448.1 | 4 - 5 | 486 | 78 | ||
XM_011523147.1 | XP_011521449.1 | 4 - 5 | 453 | 78 | ||
XM_011523148.1 | XP_011521450.1 | 4 - 5 | 526 | 78 | ||
XM_011523151.1 | XP_011521453.1 | 4 - 5 | 485 | 78 | ||
XM_017023346.1 | XP_016878835.1 | 4 - 5 | 731 | 78 | ||
GenBank mRNA | AK022933.1 | - | 4 - 5 | 493 | 78 | |
AK299016.1 | - | 4 - 5 | 448 | 78 | ||
AK299348.1 | - | 4 - 5 | 491 | 78 | ||
AK308296.1 | - | 3 - 4 | 278 | 78 | ||
BC028351.1 | - | 4 - 5 | 440 | 78 | ||
BP228881.1 | - | 4 - 5 | 478 | 78 | ||
DB079205.1 | - | 4 - 5 | 526 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
