Assay Details
Gene Symbol: LINC01229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104661.1 | 2 - 3 | 100 | 60 | ||
GenBank mRNA | BX116522.1 | - | 2 - 3 | 95 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000567665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | ENST00000569164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011435.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | NR_104660.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | TC1600011435.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | NR_104661.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | ENST00000568751.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600191598.hg.1 | NR_104660.1 | Refseq |
