Assay Details
Gene Symbol: LINC01229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104660.1 | 2 - 3 | 300 | 76 | ||
NR_104661.1 | 3 - 4 | 338 | 76 | |||
GenBank mRNA | BQ189302.1 | - | 2 - 3 | 300 | 76 | |
BX116522.1 | - | 3 - 4 | 333 | 76 | ||
CK299455.1 | - | 2 - 3 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
