Assay Details
Gene Symbol: LOC101929741 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125930.1 | 2 - 3 | 97 | 75 | ||
GenBank mRNA | BF507886.1 | - | 2 - 3 | 84 | 75 | |
BQ438340.1 | - | 2 - 3 | 98 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
