Assay Details
Gene Symbol: FOXD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121636.1 | 1 - 2 | 180 | 75 | ||
GenBank mRNA | KF925350.1 | - | 1 - 2 | 87 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | TC0100014420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207986.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | TC0100014420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207992.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207993.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207993.hg.1 | TC0100014420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207993.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
