Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027896.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027896.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027896.hg.1 |
TC05002008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027897.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027897.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05027897.hg.1 |
TC05002008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188351.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188351.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188351.hg.1 |
TC0500012661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500188351.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188351.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012661.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188350.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188350.hg.1 |
NM_031908.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500188350.hg.1 |
TC0500012661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500188350.hg.1 |
ENST00000393975.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500188350.hg.1 |
NM_031908.4 |
Refseq |