Assay Details
Gene Symbol: BOD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148894.2 | NP_683692.2 | 2 - 3 | 481 | 82 | |
XM_005248150.3 | XP_005248207.1 | 2 - 3 | 498 | 82 | ||
XM_005248151.3 | XP_005248208.1 | 2 - 3 | 498 | 82 | ||
XM_006713958.3 | XP_006714021.1 | 2 - 3 | 498 | 82 | ||
XM_011513827.2 | XP_011512129.1 | 2 - 3 | 498 | 82 | ||
XM_011513829.2 | XP_011512131.1 | 2 - 3 | 498 | 82 | ||
XM_017008009.1 | XP_016863498.1 | 2 - 3 | 498 | 82 | ||
XM_017008010.1 | XP_016863499.1 | 2 - 3 | 498 | 82 | ||
GenBank mRNA | AF528529.1 | - | 2 - 3 | 481 | 82 | |
BC065546.1 | - | 2 - 3 | 430 | 82 | ||
BC087835.1 | - | 2 - 3 | 487 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001047.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04014347.hg.1 | TC04001047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04014347.hg.1 | ENST00000040738.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04014347.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000505343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000507943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000509897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | ENST00000511119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010085.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | ENST00000040738.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | ENST00000482713.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | NM_148894.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400169137.hg.1 | TC0400010085.hg.1 | Clariom D TC |
