Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020319.2 | NP_064715.1 | 8 - 9 | 1252 | 77 | |
XM_011515458.1 | XP_011513760.1 | 9 - 10 | 1379 | 77 | ||
GenBank mRNA | AL050390.1 | - | 8 - 9 | 1194 | 77 | |
BC015453.1 | - | 2 - 3 | 170 | 77 | ||
BC035353.1 | - | 8 - 9 | 1208 | 5185501 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001161.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07018167.hg.1 | TC07001161.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018167.hg.1 | ENST00000306999.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018167.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173574.hg.1 | TC0700010352.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000306999.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000421746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000447802.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000453623.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000627859.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | ENST00000628652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010352.hg.1 | NM_020319.2 | Refseq |
