Assay Details
Gene Symbol: LTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000595.3 | NP_000586.2 | 1 - 2 | 149 | 73 | |
GenBank mRNA | BG685356.1 | - | 1 - 2 | 150 | 73 | |
D00102.1 | - | 1 - 2 | 49 | 73 | ||
D12614.1 | - | 1 - 2 | 127 | 73 | ||
X01393.1 | - | 1 - 2 | 66 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000370.hg.1, TC6_cox_hap2000066.hg.1, TC6_dbb_hap3000057.hg.1, TC6_mann_hap4000058.hg.1, TC6_mcf_hap5000052.hg.1, TC6_qbl_hap6000057.hg.1, TC6_ssto_hap7000053.hg.1
Additional Information:
This assay is in the 5'UTR region of the D00102.1, D12614.1, NM_000595.3, X01393.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of chronic inflammatory response to antigenic stimulus
positive regulation of humoral immune response mediated by circulating immunoglobulin
apoptotic process
humoral immune response
signal transduction
cell-cell signaling
response to nutrient
response to xenobiotic stimulus
response to lipopolysaccharide
positive regulation of interferon-gamma production
tumor necrosis factor-mediated signaling pathway
positive regulation of apoptotic process
negative regulation of fibroblast proliferation
lymph node development
defense response to Gram-positive bacterium
positive regulation of glial cell proliferation
signaling receptor binding
cytokine activity
tumor necrosis factor receptor binding
response to hypoxia
positive regulation of chronic inflammatory response to antigenic stimulus
positive regulation of humoral immune response mediated by circulating immunoglobulin
apoptotic process
humoral immune response
signal transduction
cell-cell signaling
response to nutrient
response to xenobiotic stimulus
response to lipopolysaccharide
positive regulation of interferon-gamma production
tumor necrosis factor-mediated signaling pathway
positive regulation of apoptotic process
negative regulation of fibroblast proliferation
lymph node development
defense response to Gram-positive bacterium
positive regulation of glial cell proliferation
signaling receptor binding
cytokine activity
tumor necrosis factor receptor binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000880.hg.1 | TC6_dbb_hap3000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000880.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000880.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000729.hg.1 | TC6_ssto_hap7000053.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000729.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000729.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000915.hg.1 | TC6_qbl_hap6000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000915.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000918.hg.1 | TC6_qbl_hap6000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000918.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000918.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000937.hg.1 | TC6_cox_hap2000066.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000937.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000937.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000890.hg.1 | TC6_mann_hap4000058.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000890.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000726.hg.1 | TC6_ssto_hap7000053.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000726.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000752.hg.1 | TC6_mcf_hap5000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000752.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000752.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000749.hg.1 | TC6_mcf_hap5000052.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000749.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003540.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003540.hg.1 | ENST00000436827.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003540.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003540.hg.1 | TC06000370.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000877.hg.1 | TC6_dbb_hap3000057.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000877.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | ENST00000436827.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | ENST00000454783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003543.hg.1 | TC06000370.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000934.hg.1 | TC6_cox_hap2000066.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000934.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000893.hg.1 | TC6_mann_hap4000058.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000893.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000893.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000454783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | ENST00000489638.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007595.hg.1 | NM_001159740.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | ENST00000471842.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | TC0600007595.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | NM_000595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | ENST00000418386.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151737.hg.1 | ENST00000471842.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 163567 | - | - |
