Assay Details
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000171.3 | NP_000162.2 | 5 - 6 | 850 | 86 | |
NM_001146040.1 | NP_001139512.1 | 5 - 6 | 850 | 86 | ||
NM_001292000.1 | NP_001278929.1 | 4 - 5 | 722 | 86 | ||
XM_017009349.1 | XP_016864838.1 | 5 - 6 | 611 | 86 | ||
GenBank mRNA | AK226046.1 | - | 4 - 5 | 722 | 86 | |
AK308539.1 | - | 5 - 6 | 846 | 86 | ||
AK312702.1 | - | 5 - 6 | 776 | 86 | ||
BC074980.2 | - | 5 - 6 | 604 | 30915298 | 86 | |
BC114947.1 | - | 5 - 6 | 850 | 40084102 | 86 | |
BC114967.1 | - | 4 - 5 | 722 | 40084103 | 86 | |
X52009.1 | - | 5 - 6 | 859 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001960.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
action potential
startle response
regulation of respiratory gaseous exchange by nervous system process
ion transport
chloride transport
muscle contraction
neuropeptide signaling pathway
acrosome reaction
visual perception
adult walking behavior
ion transmembrane transport
regulation of membrane potential
neuromuscular process controlling posture
protein homooligomerization
protein heterooligomerization
negative regulation of transmission of nerve impulse
synaptic transmission, glycinergic
righting reflex
inhibitory postsynaptic potential
cellular response to amino acid stimulus
cellular response to zinc ion
cellular response to ethanol
response to alcohol
chloride transmembrane transport
positive regulation of acrosome reaction
protein binding
zinc ion binding
glycine binding
extracellularly glycine-gated chloride channel activity
transmitter-gated ion channel activity
taurine binding
action potential
startle response
regulation of respiratory gaseous exchange by nervous system process
ion transport
chloride transport
muscle contraction
neuropeptide signaling pathway
acrosome reaction
visual perception
adult walking behavior
ion transmembrane transport
regulation of membrane potential
neuromuscular process controlling posture
protein homooligomerization
protein heterooligomerization
negative regulation of transmission of nerve impulse
synaptic transmission, glycinergic
righting reflex
inhibitory postsynaptic potential
cellular response to amino acid stimulus
cellular response to zinc ion
cellular response to ethanol
response to alcohol
chloride transmembrane transport
positive regulation of acrosome reaction
protein binding
zinc ion binding
glycine binding
extracellularly glycine-gated chloride channel activity
transmitter-gated ion channel activity
taurine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | ENST00000274576.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | ENST00000545569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05027288.hg.1 | TC05001960.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | TC0500012526.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500187446.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000274576.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000455880.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000462581.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | ENST00000471351.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_000171.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001146040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500012526.hg.1 | NM_001292000.1 | Refseq |
