Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020809.hg.1 |
ENST00000369449.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020809.hg.1 |
ENST00000465553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020809.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X020809.hg.1 |
TC0X001551.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
TC0X00011238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00176885.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000321926.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000369449.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000465553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
ENST00000491205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011238.hg.1 |
NM_001289.5 |
Refseq |