Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000279146.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000525341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000528641.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
ENST00000529797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302959.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_001302960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008139.hg.1 |
NM_003977.3 |
Refseq |