Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004198.3 | NP_004189.1 | 2 - 3 | 577 | 85 | |
GenBank mRNA | AK313521.1 | - | 2 - 3 | 423 | 85 | |
BC014456.1 | - | 2 - 3 | 267 | 4862198 | 85 | |
U62435.1 | - | 2 - 3 | 363 | 85 | ||
Y16282.1 | - | 2 - 3 | 221 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001182.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
signal transduction
chemical synaptic transmission
synaptic transmission, cholinergic
neuromuscular synaptic transmission
regulation of dopamine secretion
response to nicotine
membrane depolarization
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
transport
signal transduction
chemical synaptic transmission
synaptic transmission, cholinergic
neuromuscular synaptic transmission
regulation of dopamine secretion
response to nicotine
membrane depolarization
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08015243.hg.1 | TC08001182.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08015243.hg.1 | ENST00000276410.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08015243.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08015243.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000529467.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | ENST00000534622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_001199279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010297.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000276410.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000530869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | ENST00000533810.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | NM_004198.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800170041.hg.1 | TC0800010297.hg.1 | Clariom D TC |
