Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008169.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
ENST00000539460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
TC2200008169.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
NR_110539.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
ENST00000320372.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200158429.hg.1 |
ENST00000539460.1 |
Ensembl |