Assay Details
Gene Symbol: MRPL50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019051.2 | NP_061924.1 | 2 | 727 | 148 | |
GenBank mRNA | AK000500.1 | - | 2 | 704 | 148 | |
BC032008.1 | - | 2 | 689 | 4749199 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854834 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854831 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198255 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198257 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27198260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06854824 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 16503 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 13746 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 782294 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 179397 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 735269 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P253412 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P253412 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1608 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P120112 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P120112 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 34362 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P18714711 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P16944028 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P253412 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P253412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16248497 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198255 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198257 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854832 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854831 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198255 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16248497 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854834 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06854826 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27198257 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P253412 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P120112 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854821 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854825 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854823 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854834 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854832 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854830 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854828 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06854831 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P06854834 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16248499 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P06854833 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P06854835 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P06854836 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P120112 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P120112 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P253412 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 10734 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 73427 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18714708 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38815707 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P120112 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854823 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854832 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854809 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854811 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854810 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854799 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854819 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854804 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854821 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854813 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854825 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854834 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16248484 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854807 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854802 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854817 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854830 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854791 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854831 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854815 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854828 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854816 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16248482 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06854795 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P120112 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_019051_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162883 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162881 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162882 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8162880 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 225581_PM_s_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 243219_PM_x_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 225580_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11760538_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11721262_at | - | - |
Affymetrix Human Genome U219 Array | 11760538_a_at | - | - |
Affymetrix Human Genome U219 Array | 11721262_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900175181.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900175181.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900175181.hg.1 | TC0900011031.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900175181.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900175181.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | ENST00000374865.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011031.hg.1 | NM_019051.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 243219_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 225580_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 225581_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 243219_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 225581_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 225580_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_019051_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225580_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 225581_s_at4 | - | - |
Human Genome U95 Set | 48485_at | - | - |
Human Genome U95 Set | 48485_PM_at | - | - |
Human Genome U133 Set | 225581_PM_s_at | - | - |
Human Genome U133 Set | 225581_s_at | - | - |
Human Genome U133 Set | 243219_PM_x_at | - | - |
Human Genome U133 Set | 243219_x_at | - | - |
Human Genome U133 Set | 225580_at | - | - |
Human Genome U133 Set | 225580_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037819 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037819 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 226996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1664833 | AK000500.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21265095-S | CN388719.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21265095-S | NM_019051.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21265095-S | AK000500.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21265095-S | BC032008.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_19680-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001770131 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001770131 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7150315 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_19680 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1664833 | BC032008.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1664833 | NM_019051.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1664833 | AK000500.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1664833 | BC032008.1 | human_transcriptome |
