Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036871.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036871.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036871.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036874.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036874.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036874.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036874.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036876.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036876.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036876.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007236.hg.1 |
ENST00000481504.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036875.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036875.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036875.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036875.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036870.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036870.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036870.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036868.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036868.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036868.hg.1 |
TC02002292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203793.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203793.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203793.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203793.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203793.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203795.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203798.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007236.hg.1 |
ENST00000481504.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000463963.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000469019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014204.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203794.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203794.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203794.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203794.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203794.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203800.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203800.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203800.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203800.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203800.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
NM_004807.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
TC0200014204.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
ENST00000259241.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
ENST00000494089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203799.hg.1 |
NM_004807.2 |
Refseq |