Assay Details
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145165.1 | NP_001138637.1 | 3 - 4 | 805 | 142 | |
NM_031304.4 | NP_112594.1 | 3 - 4 | 516 | 142 | ||
GenBank mRNA | BC002817.2 | - | 3 - 4 | 516 | 3635058 | 142 |
BC009863.2 | - | 2 - 3 | 405 | 3938535 | 142 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001043.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19014214.hg.1 | ENST00000250937.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014214.hg.1 | ENST00000427575.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19014214.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014214.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19014214.hg.1 | TC19001043.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009289.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | NM_031304.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | TC1900009289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000427575.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000250937.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000586906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000587122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | ENST00000592858.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | NM_001145165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900174016.hg.1 | NM_031304.4 | Refseq |
