Assay Details
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 550 | 217 | |
GenBank mRNA | BC096732.1 | - | 1 | 543 | 40015059 | 217 |
BC098355.1 | - | 1 | 543 | 40015056 | 217 | |
M11026.1 | - | 1 | 550 | 217 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000954.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
blood coagulation
response to virus
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
cytokine activity
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
blood coagulation
response to virus
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
cytokine activity
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925890 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06637780 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16164976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925858 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06637779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26925877 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 79697 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2779 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2778 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P403459 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P20704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164952 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164952 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925886 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06637779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925858 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925885 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16164971 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26925882 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06637778 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06637779 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06637780 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637779 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637778 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06637776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_021268_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160388 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160398 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160397 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160379 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160380 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160396 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160395 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8160399 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8160377 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8160394 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 211405_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11741919_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11741919_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09012701.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09012701.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09012701.hg.1 | TC09000954.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09012702.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09012702.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09012702.hg.1 | TC09000954.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012216.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185716.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185716.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185716.hg.1 | TC0900012216.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900185716.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185716.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185717.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185717.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185717.hg.1 | TC0900012216.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900185717.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185717.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185718.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185718.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185718.hg.1 | TC0900012216.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900185718.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185718.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185719.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185719.hg.1 | NM_021268.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900185719.hg.1 | TC0900012216.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900185719.hg.1 | ENST00000413767.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900185719.hg.1 | NM_021268.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 211405_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 211405_x_at | - | - |
Human Genome U133A 2.0 Array | 211405_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 211405_x_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw503142 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 211405_x_at8 | - | - |
Human Genome U133 Set | 211405_x_at | - | - |
Human Genome U133 Set | 211405_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037180 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037180 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 104379 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1652376 | M11026.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_10880984-S | M11026.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_10880984-S | NM_021268.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_25972-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0004570520 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0004570520 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_25972 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_25972 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1652376 | NM_021068.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652376 | M11026.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652376 | NM_021268.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652376 | NM_021068.2 | human_transcriptome |
