Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156317.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156317.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156317.hg.1 |
TC0X00008243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156317.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156317.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008243.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156315.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156315.hg.1 |
NR_110396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156315.hg.1 |
TC0X00008243.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00156315.hg.1 |
ENST00000428222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156315.hg.1 |
NR_110396.1 |
Refseq |