Assay Details
Gene Symbol: CAPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145122.1 | NP_001138594.1 | 2 - 3 | 372 | 82 | |
NM_001321270.1 | NP_001308199.1 | 3 - 4 | 573 | 82 | ||
XM_011532863.2 | XP_011531165.1 | 3 - 4 | 533 | 82 | ||
XM_011532864.2 | XP_011531166.1 | 3 - 4 | 570 | 82 | ||
XM_011532865.1 | XP_011531167.1 | 3 - 4 | 474 | 82 | ||
XM_011532866.2 | XP_011531168.1 | 2 - 3 | 378 | 82 | ||
GenBank mRNA | AK092257.1 | - | 3 - 4 | 474 | 82 | |
AK296910.1 | - | 4 - 5 | 340 | 82 | ||
DC347418.1 | - | 4 - 5 | 372 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001715.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK092257.1,NM_001321270.1,XM_011532866.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02027694.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027694.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027694.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027694.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027696.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027696.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027696.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027696.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027695.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027695.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027695.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027695.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188840.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188839.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188841.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188841.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188841.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188841.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188841.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188838.hg.1 | TC0200012168.hg.1 | Clariom D TC |
