Assay Details
Gene Symbol: SLC5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003041.3 | NP_003032.1 | 5 - 6 | 592 | 75 | |
NR_130783.1 | 5 - 6 | 592 | 75 | |||
XM_006721072.3 | XP_006721135.2 | 5 - 6 | 1175 | 75 | ||
GenBank mRNA | BC131542.1 | - | 5 - 6 | 592 | 40085355 | 75 |
M95549.1 | - | 5 - 6 | 593 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000379.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
