Assay Details
Gene Symbol: LOC105373860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK026761.1 | - | 1 | 285 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | erb-b2 receptor tyrosine kinase 4 | ERBB4 |
ALS19, HER4, p180erbB4 |
-167.6kb | Sense | Chr. 2 - 211375717 - 212539147 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC102725079 | LOC102725079 | 86.5kb | Anti-sense | Chr. 2 - 212795331 - 212819263 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
