Assay Details
Gene Symbol: LOC388882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110539.1 | 9 - 10 | 792 | 105 | ||
GenBank mRNA | BC036910.1 | - | 9 - 10 | 792 | 5164270 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158427.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158427.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158427.hg.1 | TC2200008169.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158427.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158427.hg.1 | NR_110539.1 | Refseq |
