Assay Details
Gene Symbol: LAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122606.1 | NP_001116078.1 | 2 - 3 | 364 | 124 | |
NM_002294.2 | NP_002285.1 | 2 - 3 | 364 | 124 | ||
NM_013995.2 | NP_054701.1 | 2 - 3 | 364 | 124 | ||
GenBank mRNA | AK291090.1 | - | 2 - 3 | 311 | 124 | |
AK304405.1 | - | 2 - 3 | 211 | 124 | ||
AY561849.1 | - | 2 - 3 | 184 | 124 | ||
BC002965.1 | - | 2 - 3 | 319 | 3543019 | 124 | |
DA405328.1 | - | 2 - 3 | 364 | 124 | ||
J04183.1 | - | 2 - 3 | 282 | 124 | ||
S79873.1 | - | 2 - 3 | 321 | 124 | ||
U36336.1 | - | 2 - 3 | 321 | 124 | ||
X77196.1 | - | 2 - 3 | 321 | 124 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001325.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304405.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | ENST00000200639.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | ENST00000434600.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | ENST00000540603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017787.hg.1 | TC0X001325.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | TC0X00010671.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00172546.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
