Assay Details
Gene Symbol: ATAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014109.3 | NP_054828.2 | 26 - 27 | 4114 | 135 | |
XM_011516994.2 | XP_011515296.1 | 25 - 26 | 4251 | 135 | ||
XM_011516996.2 | XP_011515298.1 | 15 - 16 | 2534 | 135 | ||
GenBank mRNA | AF118088.1 | - | 9 - 10 | 1572 | 135 | |
AL833653.1 | - | 26 - 27 | 3868 | 135 | ||
AY598335.1 | - | 26 - 27 | 4006 | 135 | ||
BC010686.1 | - | 11 - 12 | 1982 | 135 | ||
BC019909.1 | - | 8 - 9 | 1318 | 135 | ||
BC113656.1 | - | 26 - 27 | 4028 | 8322708 | 135 | |
CR749832.1 | - | 27 - 28 | 4343 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001584.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
