Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188041.hg.1 |
TC0800012455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012455.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188045.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188045.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188045.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188045.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188045.hg.1 |
TC0800012455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
ENST00000357082.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
ENST00000622816.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
NM_001204180.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188043.hg.1 |
TC0800012455.hg.1 |
Clariom D TC |