Assay Details
Gene Symbol: ZHX1-C8orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204180.1 | NP_001191109.1 | 3 - 4 | 487 | 110 | |
GenBank mRNA | BC012379.2 | - | 2 - 3 | 217 | 3857253 | 110 |
BP340117.1 | - | 3 - 4 | 285 | 110 |
Gene Symbol: C8orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032847.2 | NP_116236.1 | 2 - 3 | 271 | 110 | |
GenBank mRNA | AK027731.1 | - | 2 - 3 | 250 | 110 | |
AK222660.1 | - | 2 - 3 | 219 | 110 | ||
BC067796.1 | - | 2 - 3 | 245 | 5260773 | 110 | |
DR156993.1 | - | 2 - 3 | 271 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08002618.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08020714.hg.1 | TC08002618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08020714.hg.1 | ENST00000276704.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020714.hg.1 | ENST00000521310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020714.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08020715.hg.1 | TC08002618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08020715.hg.1 | ENST00000276704.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020715.hg.1 | ENST00000521310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020715.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08020716.hg.1 | TC08002618.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08020716.hg.1 | ENST00000276704.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020716.hg.1 | ENST00000521310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08020716.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187995.hg.1 | TC0800012453.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800188039.hg.1 | TC0800012455.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012455.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | ENST00000357082.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | ENST00000622816.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | NM_001204180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800188040.hg.1 | TC0800012455.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187994.hg.1 | TC0800012453.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800187997.hg.1 | TC0800012453.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000276704.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000517760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000518996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000519791.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000521310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000522477.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | ENST00000523726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012453.hg.1 | NM_032847.2 | Refseq |
