Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009578.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009578.hg.1 |
TC04000663.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400160115.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400160115.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400160115.hg.1 |
TC0400008725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400160115.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400160115.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000264360.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000511112.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008725.hg.1 |
NM_032961.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400160116.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400160116.hg.1 |
NM_020815.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400160116.hg.1 |
TC0400008725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400160116.hg.1 |
ENST00000618019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400160116.hg.1 |
NM_020815.2 |
Refseq |